VWR®, Desiccator Plates, Large
Supplier: VWR Collection
89038-054
VWRU89038-054EA 131 SGD
VWRU89038-054
VWR®, Desiccator Plates, Large
Desiccators Desiccators, Plates
Porcelain, glazed on one side.
  • Features eight holes with 30 mm Ø, does not have a central hole
  • Without feet
Order Now
SPECIFICATIONS
Environmentally Preferable