VWR®, Desiccator Plates
Supplier: VWR Collection
89038-068 89038-070 89038-072
VWRU89038-068EA 86.5 SGD
VWRU89038-068 VWRU89038-070 VWRU89038-072
VWR®, Desiccator Plates
Desiccators Desiccators, Plates
Porcelain plates, glazed on one side. The plates contain numerous holes of 5 mm Ø, (VWRU89038-074 has 2 mm Ø) and a large centre hole.
  • Without feet
  • Resistant to chemicals
  • Withstand maximum temperature of 1150 °C