VWR®, Filter Funnels according to Dr. Hirsch
Supplier: VWR Collection
89038-136 89038-140 89038-142
VWRU89038-136EA 47 SGD
VWRU89038-136 VWRU89038-140 VWRU89038-142
VWR®, Filter Funnels according to Dr. Hirsch
Funnels Filtration Funnels Filter Funnels
Porcelain, glased inside and out, except rim.
Funnel with integral perforated filter plate can be used with paper or membrane discs.
Order Now
SPECIFICATIONS
Environmentally Preferable