VWR®, Bunsen Burner
Supplier: VWR Collection
89038-528
VWRU89038-528EA 61.6 SGD
VWRU89038-528
VWR®, Bunsen Burner
Heaters Gas Burners
Bunsen burner with plated die-cast alloy base. Burner has 11 mm aluminium mixing tubes and 8 mm serrated gas inlet connections.
Height: 137 mm
Order Now
SPECIFICATIONS
Environmentally Preferable