VWR®, Bunsen Burner
Supplier: VWR Collection
89038-530
VWRU89038-530EA 80.5 SGD
VWRU89038-530
VWR®, Bunsen Burner
Heaters Gas Burners
Bunsen burner with nickel plated zinc alloy die-cast base.
  • Designed with a flame stabiliser to provide a steady flame for general laboratory needs
  • With aluminium mixing tubes and serrated gas inlet and air regulator
  • Optional wing top produces a broad flat flame for working glass and general lab use
Height: 156 mm
Ordering information: Wing top must be ordered separately.