VWR® Signature™, Disposable electroporation cuvettes
Supplier: VWR Collection
89047-208
VWRU89047-208EA 471 SGD
VWRU89047-208
VWR® Signature™, Disposable electroporation cuvettes
Cuvettes Electroporation Cuvettes
High quality durable polycarbonate has been tested to optimise transformation efficiencies. These cuvettes are built to provide exact tolerances ensuring reproducible results.
  • Polished aluminum plates reducing static and arcing
  • Compatible with most electroporation devices
  • Gamma irradiated to warranty sterility
Packaging: 50 individually wrapped cuvettes.