VWR®, Aluminium Foil Seals for Cold Storage
Supplier: VWR Collection
89049-034
VWRU89049-034EA 184 SGD
VWRU89049-034
VWR®, Aluminium Foil Seals for Cold Storage
Seals Microplate Seals
Aluminium foils with biologically inert adhesive. These foils are easily pierced by manual or automated pipette tips, for sample recovery without generation of aerosols or cross-contamination.
  • Recommended for temperatures from −80 to +130 °C
  • Not recommended for PCR
Each foil L×D: 133×82 mm