VWR®, Microscope Slides for Printers
Supplier: VWR Collection
89078-508
VWRU89078-508EA 0 SGD
VWRU89078-508
VWR®, Microscope Slides for Printers
Slides Microscope Slides
Slides with frosted tab designed to be printed on by inkjet printer for histology, cytology, hematology, and microbiology applications.
Delivery information: Frosted tabs are available in various colours.