VWR®, Vials, Microwave Reaction
Supplier: VWR Collection
89079-404 89079-406
VWRU89079-404EA 1500 SGD
VWRU89079-404 VWRU89079-406
VWR®, Vials, Microwave Reaction
Vials
Reaction vials made from borosilicate glass, class A, type I are designed to withstand pressures up to 21 bar.
Delivery information: Supplied with 20 mm aluminium seals with installed blue PTFE-faced silicone septa with 3,2 mm thickness.