VWR®, Centrifuge Tubes, High Performance
Supplier: VWR Collection
89039-664 89039-656 89079-494
VWRU89039-664EA 255 SGD
VWRU89039-664 VWRU89039-656 VWRU89079-494
VWR®, Centrifuge Tubes, High Performance
Tubes Centrifuge Tubes
PP, conical, with purple HDPE flat cap and integral sealing or plug-style cap and deep sealing area.
  • Certified non cytotoxic, nuclease- and pyrogen-free
  • Caps available in two styles: flat cap with integral sealing and a smooth surface for additional labelling options, or plug-style cap with deep sealing area for a secure seal when using shakers and horizontal incubators
  • Easy to read black graduations and white writing area on the side for permanent coding
  • Can be used for storing and freezing samples at temperatures down to –80 °C
  • Can be centrifuged up
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M