VWR® Ultra High Performance Centrifuge Tubes, Light-Sensitive
Supplier: VWR Collection
89079-532
VWRU89079-532EA 383 SGD
VWRU89079-532
VWR® Ultra High Performance Centrifuge Tubes, Light-Sensitive
Tubes Centrifuge Tubes
PP, conical, amber, with purple HDPE plug-style cap with deep sealing area.
  • Printed labelling area for quick and convenient sample identification
  • Available in 15 or 50 ml sizes and in a variety of packaging formats
  • Certified non cytotoxic, nuclease- and pyrogen-free
  • Sustainable design based on virgin resins
Designed to provide protection for light-sensitive samples.
Certifications: Manufactured in a class 100000 room environment. Meet the requirements of RoHS.
Ordering information: HDPE caps are included.
Order Now
SPECIFICATIONS
Environmentally Preferable