VWR®, Cryoboxes, PP
Supplier: VWR Collection
89084-934
VWRU89084-934EA 19.2 SGD
VWRU89084-934
VWR®, Cryoboxes, PP
Boxes Cryoboxes
PP, coloured.
  • Durable and autoclavable
  • An alphanumeric grid on the lid aids in sample identification
These boxes maintain their integrity under harsh freezer conditions and will not absorb water or degrade like cardboard storage boxes. A three-point hinge and positive catch provide quick access to samples and keep them secure when box is closed. Boxes are available in six colours; black is ideal for storage of light-sensitive samples.