VWR®TraceClean®, Bottles, narrow neck, round, boston type
Supplier: VWR Collection
89093-918 89093-912 89093-916
VWRU89093-918EA 254 SGD
VWRU89093-918 VWRU89093-912 VWRU89093-916
VWR®TraceClean®, Bottles, narrow neck, round, boston type
Bottles Glass Bottles
Soda lime glass, amber.
  • Amber glass is recommended for UV light-sensitive compounds
  • QA bottles include a Certificate of Analysis in every case
  • QA bottles also include bar codes with lot number and individual container number for complete traceability
Quality-Assured® (QA) bottles follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05 A 'Specifications and Guidance for Contaminant-Free Sample Containers'. Solid-top caps feature fluoropolymer resin liner and are for semi-volatiles, metals, pesticides, and PCB analyses; and select inorganic compounds.