VWR®TraceClean®, Water sampling bottles, glass, wide neck, straight
Supplier: VWR Collection
89093-984 89093-970 89093-986 89093-988 89093-940
VWRU89093-984EA 221 SGD
VWRU89093-984 VWRU89093-970 VWRU89093-986 VWRU89093-988 VWRU89093-940
VWR®TraceClean®, Water sampling bottles, glass, wide neck, straight
Bottles Water Sampling Bottles
Type III (soda-lime) glass.
  • Available with solid-top or open-top caps
  • Solid-top caps with fluoropolymer resin liner are for use in semi-volatiles, pesticides, PCBs, and phenolics analyses
  • Open-top caps with ultrasonically bonded fluoropolymer resin/silicone septa are for use in volatile organic analysis
  • Ideal for collection of soil, sediment, and slurry matrices
Quality-Assured® (QA) packers follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA packers include aMore Product Information