VWR®TraceClean®, Water sampling bottles, hdpe, cylindrical
Supplier: VWR Collection
89094-092
VWRU89094-092EA 123 SGD
VWRU89094-092
VWR®TraceClean®, Water sampling bottles, hdpe, cylindrical
Bottles Water Sampling Bottles
HDPE, translucent.
  • Available with poly-foam lined caps
  • For use in metals and multiple inorganic analyses.
Processed (PC) and Quality-Assured® (QA) bottles follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers.' QA packers include a certificate of analysis in every case. QA packers also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) packers do not receive the specialty cleaning process. All cases areMore Product Information