VWR®TraceClean®, Water sampling bottles, hdpe, wide neck
Supplier: VWR Collection
89094-110 89094-102 89094-108
VWRU89094-110EA 153 SGD
VWRU89094-110 VWRU89094-102 VWRU89094-108
VWR®TraceClean®, Water sampling bottles, hdpe, wide neck
Bottles Water Sampling Bottles
HDPE, translucent.
  • Available with poly-foam lined caps
  • For use in collecting clays, slurries, and sediment matrices for use in geotechnical, environmental, toxicity characteristic leaching procedure (TCLP) metals, inorganic compounds, and benthic organism studies
Processed (PC) and Quality-Assured® (QA) bottles follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers.' QA packers include a certificate of analysis in every case. QA packers also include bar codes with lotMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable