VWR® TraceClean®, Water sampling bottles, hdpe, wide neck
Supplier: VWR Collection
89094-130 89094-132 89094-134
VWRU89094-130EA 142 SGD
VWRU89094-130 VWRU89094-132 VWRU89094-134
VWR® TraceClean®, Water sampling bottles, hdpe, wide neck
Bottles Water Sampling Bottles
Bottles are used in collection of water samples for metals analyses and a variety of inorganic analytes including cyanide, sulphide, sulphate, chloride, and ion chromatography.
  • HDPE construction
  • Wide-mouthed design
Processed (PC) and Quality-Assured® (QA) packers follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A "Specifications and Guidance for Contaminant-Free Sample Containers." QA packers include a certificate of analysis in every case. QA packers also include bar codes with lot number and individual container number forMore Product Information