VWR® Reagent Reservoirs
Supplier: VWR Collection
89094-670 89094-658 89094-662
VWRU89094-670EA 264 SGD
VWRU89094-670 VWRU89094-658 VWRI613-1172 VWRI613-1173 VWRI613-1186 VWRI613-1174 VWRI613-1175 VWRI613-1176 VWRI613-1177 VWRI613-1178 VWRI613-1179 VWRI613-1180 VWRI613-1182 VWRI613-1183 VWRI613-1171 VWRI613-1185 VWRI613-1187 VWRI613-1184 VWRU89094-662
VWR® Reagent Reservoirs
Reagent Reservoirs
Designed for use in multi-channel pipetting applications, these disposable reservoirs accommodate all types of pipettes up to 12-channel width.
  • Trough design, graduation marks on interior walls, and pour-off spouts on each corner
  • Lot certified, non pyrogenic, DNase- and RNase-free
  • Non sterile or electron beam sterilised
Ordering information: 25 ml reservoirs also available in a unique divided format that features three distinct mini reservoir chambers, each with a 5 ml capacity; 50 ml reservoirs available in clear FDA-grade PVC or virgin-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable