VWR®, Reagent reservoirs
Supplier: VWR Collection
89094-682
VWRU89094-682EA 280 SGD
VWRU89094-682
VWR®, Reagent reservoirs
Reagent Reservoirs
PS, sterile or non sterile. These disposable pipetting reservoirs are designed for use in multi channel pipetting applications, and will accommodate all types of pipettors up to 12-channel width.
  • Lot-certified, non pyrogenic, DNase- and RNase-free
  • Trough design, graduation marks on interior walls, and pour-off spouts on each corner
  • 25 ml reservoirs are also available in a unique divided format that features three distinct mini reservoir chambers, each with a 5 ml capacity
Packaging: Available bulk packed or individually wrapped.