VWR®, General purpose PFA-coated thermometers
Supplier: VWR Collection
89095-658
VWRU89095-658EA 30 SGD
VWRU89095-658
VWR®, General purpose PFA-coated thermometers
Thermometers Analogue Thermometers
These mercury free thermometers are ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, manufacturing, dairies, breweries and distilleries. Blue organic liquid.
  • PFA coating keeps liquid encapsulated in the event of breakage
  • Lead-free white back glass
  • Individually serialised
Certifications: Traceable to NIST.
Delivery information: Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) statement of accuracy and traceability.