VWR®, Rubber Stoppers
Supplier: VWR Collection
89097-548 89097-554 89097-544
VWRU89097-548EA 95.5 SGD
VWRU89097-548 VWRU89097-554 VWRU89097-544
VWR®, Rubber Stoppers
Stoppers Rubber Stoppers
These stoppers allow insertion of a syringe needle and exhibit excellent resealing properties for air sensitive chemistry.
  • Natural rubber
  • Hollow plug to fit inside bottles, flasks, vials or tubing
  • Sleeves to pull over neck of bottle to form a double seal
Order Now
SPECIFICATIONS
Environmentally Preferable