VWR®, Red Border Tape
Supplier: VWR Collection
89098-108
VWRU89098-108EA 13 SGD
VWRU89098-108
VWR®, Red Border Tape
Labels
Tape labels are designed to aid in the identification of containers, shelves and other standard laboratory or workplace items.
  • Self-adhesive
  • Removes cleanly without residues
  • White with red border
  • Labels can be written on with pen, pencil or permanent marker
Delivery information: Labels are delivered on individual rolls each containing 12,7 m of tape.