VWR®, Sample container with snap-on lids
Supplier: VWR Collection
89125-846
VWRU89125-846EA 266 SGD
VWRU89125-846
VWR®, Sample container with snap-on lids
Sample Containers General Purpose Containers
PP, transparent, graduated.
  • Shatter resistant
  • Snap-on lid creates secure seal
  • Can also be sterilised
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904