VWR®, Biohazard Bags, PP
Supplier: VWR Collection
14220-068 14220-046 89129-468 89130-796 89129-470 89129-472
VWRI129-0025EA 383 SGD
VWRI129-0025 VWRU14220-068 VWRU14220-046 VWRU89129-468 VWRU89130-796 VWRU89129-470 VWRU89129-472
VWR®, Biohazard Bags, PP
Bags Waste Disposal Bags
Sturdy bags, ideal for disposing of biologically hazardous waste that must first be autoclaved. Complete with wire closures that allow expansion of the bag and ventilation when cooling. Highly resistant to tears and punctures. Available as transparent bags without temperature indicator, or orange with temperature indicator. PP, 50 µm thick, with 'Biohazard' label.
  • Autoclavable up to 135 °C
  • The useful temperature indicator text is white before autoclaving, then black
  • 'Contents Have Been Autoclaved' indicates successful autoclaving
  • Highly visible '
More Product Information