VWR®, Serological Pipettes, Disposable
Supplier: VWR Collection
89130-900
VWRU89130-900EA 3340 SGD
VWRU89130-900
VWR®, Serological Pipettes, Disposable
Pipettes Serological Pipettes
Pipettes made from PS for sterile serological and tissue culture applications.
  • Calibrated to deliver
  • Easy-to-read black ascending and descending graduations simplify fluid measuring and dispensing
  • All pipettes feature negative graduations for extra working volume
  • Pipettes are colour-coded for quick size identification
  • Accuracy is ±2% at full volume
Certifications: All pipettes are in compliance with ASTM Standard E-934, Standard Specification for Serological Pipettes, Disposable Plastic.
Packaging: Available in different packaging options:More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable