VWR®, UV Handheld Lamps
Supplier: VWR Collection
89131-498
VWRU89131-498EA 1970 SGD
VWRU89131-498
VWR®, UV Handheld Lamps
Lights UV Lights, Lamps and Bulbs
These UV lamps have many uses depending on the wavelength of emitted UV light.
  • Dual UV lamps
  • Use 6-watt tubes and are manufactured of lightweight rugged aluminum with a scratch-resistant powder paint
  • UV-A hand lamp features two 365 nm longwave UV tubes for brilliant fluorescence
Applications for 365 nm UV include water testing, quality control inspection, non destructive testing and sanitation. The 302 nm UV can be used in UV curing, gel viewing, and mineralogy.