VWR®, Spill response kit, HazMat
Supplier: VWR Collection
89168-998
VWRU89168-998EA 192 SGD
VWRU89168-998
VWR®, Spill response kit, HazMat
Spill Control Materials
This portable kit handles all types of lab spills.
Certifications: Supports compliance with 40 CFR 263.30(a) safety standard.
Delivery information: Kit contains twenty pads, three socks, one pair nitrile gloves, one pair safety goggles, and one water-resistant yellow bag with tie.
Order Now
SPECIFICATIONS
Environmentally Preferable