VWR®, Racks for Centrifuge Tubes 15 and 50 ml
Supplier: VWR Collection
89214-722
VWRU89214-722EA 618 SGD
VWRU89214-722
VWR®, Racks for Centrifuge Tubes 15 and 50 ml
Racks Tube Racks
Durable stainless steel freezer racks facilitate organisation and maximise valuable storage space.
  • Racks accommodate 15 ml and 50 ml centrifuge tubes
  • Reorder by simply scanning the QR code with smartphone or tablet
  • Sturdy handles
  • Stainless steel construction
Racks feature sturdy handles and come standard with identification labels for inventory purposes.