VWR® CryoPro® , Plastic storage boxes
Supplier: VWR Collection
89214-760
VWRU89214-760EA 25.4 SGD
VWRU89214-760
VWR® CryoPro® , Plastic storage boxes
Boxes Cryoboxes
Polycarbonate construction for use in mechanical and LN₂ freezers.
  • Grid format for lids
  • Boxes feature drain slots for safe liquid nitrogen storage
Lids come printed with 81 and 100 cell grids to assist with inventory control.