VWR®, Petri Dishes, Disposable
Supplier: VWR Collection
25384-168 25384-094 25384-322 25384-308 25384-302 89107-632 25384-088 25384-342 89230-472 25384-090 25384-164
VWRU25384-168EA 269 SGD
VWRU25384-168 VWRU25384-094 VWRU25384-322 VWRU25384-308 VWRU25384-302 VWRU89107-632 VWRU25384-088 VWRU25384-342 VWRU89230-472 VWRU25384-090 VWRU25384-164
VWR®, Petri Dishes, Disposable
Culture Dishes Petri Dishes
High quality virgin PS, clear for optimum flatness and high optical clarity. Ideal for microbial culture, where accurate bacterial count or microscopic examination is required.
  • Distortion-free visibility
  • Available in varied sizes and styles for specialised and routine applications
  • Stacking ring and slip-ring features available
VWR® Petri dishes are available in a variety of sizes and styles to accommodate both specialised and routine needs. The stacking ring feature, on some models, facilitates positive stacking while the slip-ring feature, on otherMore Product Information