VWR®, Stackable Freezer Boxes
Supplier: VWR Collection
89236-008 89235-996 89236-002
VWRU89236-008EA 84.8 SGD
VWRU89236-008 VWRU89235-996 VWRU89236-002
VWR®, Stackable Freezer Boxes
Boxes Cryoboxes
VWR® Stackable Freezer Boxes are suitable to store in temperatures as low as –86 °C.
  • Made with PP material
  • Base made with colour options natural, blue, green, purple, and orange
  • Lid has grid option for coding
Boxes are stackable. Designed for 50, 81, or 100 places to store both internal and external threaded vials (1 to 2 ml), and has slot width of 12,5 mm (±0,3 mm).