VWR®, Crucible Tongs, Stainless steel, with Corrugated Tip
Supplier: VWR Collection
89259-892
VWRU89259-892EA 64.3 SGD
VWRU89259-892
VWR®, Crucible Tongs, Stainless steel, with Corrugated Tip
Tongs
Stainless steel. These crucible holders have tight-fitting corrugated tips for handling dishes in muffle furnaces.
Constructed from a stainless steel rod with a diameter of 9 mm. There is a joint near the middle for holding large-sized crucibles.