VWR®, Precision tweezers
Supplier: VWR Collection
89259-984
VWRU89259-984EA 160 SGD
VWRU89259-984
VWR®, Precision tweezers
Forceps
These precision tweezers are made from stainless steel and feature very sharp, fine points.
  • Anti-magnetic properties
  • Resistant to acids
Order Now
SPECIFICATIONS
Environmentally Preferable