VWR®, Ovens with Forced Convection
Supplier: VWR Collection
89511-414
VWRU89511-414EA 10610 SGD
VWRU89511-414
VWR®, Ovens with Forced Convection
Ovens Standard Ovens
Forced air ovens are the optimal choice for fast drying processes, removing humidity much faster than gravity convection units. The technology ensures great temperature uniformity and stability throughout the chamber, providing ideal conditions for material testing and heat treatments. Air exchange can be controlled by a damper which can be opened for drying to remove humidity quickly and closed for heat treatment with highest requirements for temperature accuracy.
  • Consistent results with outstanding temperature stability and uniformity
  • Versatile, covering
More Product Information