VWR®, Serological Pipettes, Disposable
Supplier: VWR Collection
93000-694 93000-702 93000-704 93000-706 93000-742 93000-674 93000-680 93000-696 93000-698 93000-682 93000-678
VWRU93000-694EA 531 SGD
VWRU93000-694 VWRU93000-702 VWRU93000-704 VWRU93000-706 VWRU93000-742 VWRU93000-674 VWRU93000-680 VWRU93000-696 VWRU93000-698 VWRU93000-682 VWRU93000-678
VWR®, Serological Pipettes, Disposable
Pipettes Serological Pipettes
Borosilicate glass, Type I, Class B.
  • Uniform pulled tip, calibrated to deliver
  • Negative graduations increase versatility
  • Non sterile, non plugged pipettes feature a blue scale
These disposable serological pipettes are available in sterile or non sterile formats, with a variety of packaging options to choose from. With two rings near the top denoting that the small amount remaining in the tip after free delivery has ceased must be blown out and added to the main volume delivered.
Packaging: Individually wrapped pipettes are packaged in colour-codedMore Product Information