VWR®, Urine collection containers
Supplier: VWR Collection
95037-276
VWRU95037-276EA 300 SGD
VWRU95037-276
VWR®, Urine collection containers
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
HDPE, amber.
  • Leak resistant closure system
  • Molded graduations provide volume determination
  • EZ-UP pour spout for easy pouring out
The containers available in both horizontal or vertical space-saving format. The ergonomic handle provides convenient and balanced handling when the container is empty or filled. The containers can be disposed of using an incinerator system.