VWR®, Autoclavable Bags, PP
Supplier: VWR Collection
95042-552 95042-558 95042-554
VWRU95042-552EA 117 SGD
VWRU95042-552 VWRU95042-558 VWRU95042-554
VWR®, Autoclavable Bags, PP
Bags Waste Disposal Bags
These sturdy, transparent 50 µm thick PP bags are suitable for autoclaving hazardous and nonhazardous materials and waste.
  • Highly resistant to tears and impact
  • Temperature resistant bags and autoclavable up to 135 °C
  • Twist ties allow proper closure for bag expansion and venting during sterilisation
Sturdy, temperature resistant bags, ideal for autoclaving hazardous, nonhazardous materials and waste. Supplied with twist ties that allow proper closure for bag expansion and venting during sterilisation.
Certifications: Meets strength requirements inMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable