VWR®, Embedding compound, water- soluble
Supplier: VWR Collection
95057-838
VWRU95057-838EA 40.2 SGD
VWRU95057-838
VWR®, Embedding compound, water- soluble
Stains Microscopy and staining reagents
Frozen section compound is a fast-acting, water-soluble embedding compound designed for use in frozen sectioning. The compound bonds and encapsulates tissue specimens to the object holder for cryosectioning. At a working temperature of –20 °C, it provides optimal sectioning consistency with minimal section curling.
  • Highly recommended for surgical pathology laboratories
  • Available clear or in a light blue format
  • Each bottle features an extended length tip for increased accuracy in dispensing
  • Size: 118 ml