VWR®, Absorbent Pads
Supplier: VWR Collection
95057-860
VWRU95057-860EA 586 SGD
VWRU95057-860
VWR®, Absorbent Pads
Spill Control Materials
Designed to soak up liquids, oils, and common solvents, absorbent pads provide an excellent alternative to paper towels and gauze.
  • Strong, highly absorbent, and lint-free
  • Efficient alternative to paper towels and gauze
Pads exhibit a cloth-like softness with no loose threads or fibers, making them ideal for use in coverslipping, slide preparation, and general laboratory cleanup.