VWR®, Two-Sided Disposable Cuvettes
Supplier: VWR Collection
97000-586 97000-590 97000-588
VWRU97000-586EA 175 SGD
VWRU97000-586 VWRU97000-590 VWRU97000-588
VWR®, Two-Sided Disposable Cuvettes
Cuvettes Spectrophotometer Cuvettes
These cuvettes provide accurate and reliable results. PS cuvettes ensure low variation in extinction coefficient.
  • Arrow mark to indicate direction of transmission and reduce variation
  • Macro and semi-macro sizes accommodate most sample volumes
  • In low-dust and low-scratch material
Suitable for most spectrophotometry applications in the visible and near-UV range.
PS cuvettes are suitable for routine assays between 340 and 900 nm. They ensure low variation in extinction coefficient.
Packaging: In low-dust and low-scratch material.
Order Now
SPECIFICATIONS
Environmentally Preferable