VWR® CryoGuard, Cryo Gloves
Supplier: VWR Collection
97008-214 97008-248 97008-208 97008-204 97008-206 97008-196 97008-238 97008-216 97008-210 97008-236 97008-240 97008-242 97008-228 97008-230
VWRU97008-214EA 303 SGD
VWRU97008-214 VWRU97008-248 VWRU97008-208 VWRU97008-204 VWRU97008-206 VWRU97008-196 VWRU97008-238 VWRU97008-216 VWRU97008-210 VWRU97008-236 VWRU97008-240 VWRU97008-242 VWRU97008-228 VWRU97008-230
VWR® CryoGuard, Cryo Gloves
Gloves
These multi-layer, insulated gloves are designed to protect hands and arms from the hazards of working with cryogenic materials. They can be worn for extended periods of time, as they are extremely warm, even in temperatures as low as –196 °C.
  • Lightweight, flexible, and durable
  • Inner lining wicks moisture from the skin
  • All wrist-length gloves also incorporate an uninsulated knit cuff
Water-resistant and waterproof gloves feature two layers of insulation material bonded together at the edges to trap insulating air without adding additional bulk and anMore Product Information
Order Now
SPECIFICATIONS
Model CryoGuard
Environmentally Preferable