VWR®, Guard Rail Carts
Supplier: VWR Collection
97017-712 97017-714
VWRU97017-712EA 2110 SGD
VWRU97017-712 VWRU97017-714
VWR®, Guard Rail Carts
Trolleys
Guard rails help preventing in slipping off shelves, eliminating spills and breakage.
  • Durable
  • Easy to sanitise
The presence of standard handle and leg bumpers helps in protecting cart, walls, and doorways from damage. Equipped with two or three shelves, all-swivel casters, and integrated push handle for easy maneuverability.
Capacity is the total cart capacity across all of the shelves with the weight evenly dispersed.
Stainless steel fabrication.