VWR® Personal Sized Incubator
Supplier: VWR Collection
97025-632
VWRU97025-632EA 1600 SGD
VWRU97025-632
VWR® Personal Sized Incubator
Incubators Compact Benchtop Incubators
These personal sized incubators are ideal for use in microbiology or haematology.
  • Compact size
  • Economical
  • Metal construction
The housing and door frame are entirely metal. A Plexiglas window in the door provides full visibility into the corrosion-resistant interior chamber. The included shelf can be adjusted to three different levels. Heating elements are incorporated into the housing bottom and sides to ensure temperature stability and uniformity. The top of the unit features a grommet hole that accommodates a thermometer for an accurate temperatureMore Product Information