Laboratory mixers
Supplier: WARING BLENDOR
8011ES LB20EG
WARI8011ESEA 2030 SGD
WARI8011ES WARILB20EG
Laboratory mixers
Homogenisers Standard Laboratory Blenders
Reliable devices with powerful motors. For thorough mixing, dispersion and emulsification. Special cutter heads and high speed operation make these laboratory mixers suitable for many applications. Two basic devices with capacities of 1 or 2 L and a variety of stainless steel, heat resistant glass or PC beakers are available.
Delivery information: Supplied with motor, cutter head, beaker and lid.
Order Now
SPECIFICATIONS
Environmentally Preferable