Grade 2 qualitative filter papers, Whatman™
Supplier: Whatman products (Cytiva)
1002-150
WHAT1002-150EA 65.9 SGD
WHAT1002-150
Grade 2 qualitative filter papers, Whatman™
Filters Filter Papers Qualitative Filter Paper
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
  • Each lot is guaranteed to meet the ASTM E 2187-09 specifications
  • Simplifies testing process by removing lot suitability testing
  • Just condition and use
Slightly more retentive than Grade 1 with a corresponding increase in filtration time (i.e., slightly slower filtration speed). More absorbent thanMore Product Information