Grade 3 qualitative filter papers, Whatman™
Supplier: Whatman products (Cytiva)
1003-185
WHAT1003-185EA 247 SGD
WHAT1003-185
Grade 3 qualitative filter papers, Whatman™
Filters Filter Papers Qualitative Filter Paper
A standard grade filter used in qualitative analytical techniques to determine and identify materials. For use in büchner funnels.
  • Thickness of 6 µm
Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
Double the thickness of Grade 1 with still finer particle retention and excellent loading capacity; more precipitate can be held without clogging. The extra thickness gives increased wet strength and makes this grade highly suitable for use in BüchnerMore Product Information