Grade 5 qualitative filter papers, Whatman™
Supplier: Whatman products (Cytiva)
1005-055
WHAT1005-055EA 52.5 SGD
WHAT1005-055
Grade 5 qualitative filter papers, Whatman™
Filters Filter Papers Qualitative Filter Paper
A standard grade clarifying filter excellent for cloudy suspensions and for water and soil analysis.
  • Thickness of 2,5 µm
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
The maximum degree of fine particle filtration in the qualitative range. Capable of retaining the fine precipitates encountered in chemical analysis. Slow flow rate. ExcellentMore Product Information