Grade 602 h ½ qualitative filter papers, Whatman™
Supplier: Whatman products (Cytiva)
10312651
WHAT10312651EA 0 SGD
WHAT10312651
Grade 602 h ½ qualitative filter papers, Whatman™
Filters Filter Papers Qualitative Filter Paper
A dense prepleated filter paper for collecting very small particles and removing fine precipitates.
  • Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
  • Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
  • Increased total loading capacity as more filter area is available
  • Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of
More Product Information