Extraction thimbles
Supplier: Whatman products (Cytiva)
10350324
WHAT10350324EA 0 SGD
WHAT10350324
Extraction thimbles
Extractors Extraction Thimbles
Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.