Blotting membranes, Nytran® N, Whatman™
Supplier: Whatman products (Cytiva)
10416096
WHAT10416096EA 0 SGD
WHAT10416096
Blotting membranes, Nytran® N, Whatman™
Transfer Membranes
The Nytran® Nylon (Nytran® N) membrane is ideal for applications that require a lower charge. It is designed for Southern and Northern blotting as well as colony and plaque lifts and Dot or Slot Blots. Nytran® N is compatible with isotopic detection methods.
  • Excellent signal-to-noise ratios
  • Membrane is cast uniformly on both sides of the support matrix, providing excellent symmetry and allowing the membrane to lie flat without curling
  • Uniform pore size and distribution
  • Available in 0,2 and 0,45 µm pore sizes for optimal retention of oligos and larger DNA
More Product Information